Ethyl-1-(trifluoromethyl)cyclopropanecarboxylate 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC51399-1G
APOSPC51399-1GEA 81.8 EUR
APOSPC51399-1G
Ethyl-1-(trifluoromethyl)cyclopropanecarboxylate 95%
Ethyl-1-(trifluoromethyl)cyclopropanecarboxylate
Formule: C₇H₉F₃O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD22199506
CAS nummer: 139229-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR