3-(Difluoromethyl)-2-(2,3,4-trimethoxyphenyl)-1H-indole

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC520098-1G
APOSPC520098-1GEA 2760 EUR
APOSPC520098-1G
3-(Difluoromethyl)-2-(2,3,4-trimethoxyphenyl)-1H-indole
3-(Difluoromethyl)-2-(2,3,4-trimethoxyphenyl)-1H-indole
Formule: C₁₈H₁₇F₂NO₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD29767581
CAS nummer: 2149597-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR