1-Bromo-4-[(trifluoromethyl)sulphonyl]benzene 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC520507-10G PC520507-1G PC520507-5G
APOSPC520507-10GEA 1220 EUR
APOSPC520507-10G APOSPC520507-1G APOSPC520507-5G
1-Bromo-4-[(trifluoromethyl)sulphonyl]benzene 96%
1-Bromo-4-[(trifluoromethyl)sulphonyl]benzene
Formule: C₇H₄BrF₃O₂S
Smeltpunt: 60…65 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00474580
CAS nummer: 312-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR