2-Chloro-5-fluoropyridine-3-carbaldehyde 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC520523-5G
APOSPC520523-5GEA 182 EUR
APOSPC520523-5G
2-Chloro-5-fluoropyridine-3-carbaldehyde 97%
2-Chloro-5-fluoropyridine-3-carbaldehyde
Formule: C₆H₃ClFNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03095247
CAS nummer: 851484-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR