2,4-Difluoro-3-nitrobenzonitrile 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC520622-1G PC520622-5G
APOSPC520622-1GEA 165 EUR
APOSPC520622-1G APOSPC520622-5G
2,4-Difluoro-3-nitrobenzonitrile 97%
2,4-Difluoro-3-nitrobenzonitrile
Formule: C₇H₂F₂N₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11226536
CAS nummer: 1186194-75-1
UN: 3276
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR