1-(1,1-Difluoroethyl)-4-fluorobenzene 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC520666-1G PC520666-500MG PC520666-5G
APOSPC520666-1GEA 317 EUR
APOSPC520666-1G APOSPC520666-500MG APOSPC520666-5G
1-(1,1-Difluoroethyl)-4-fluorobenzene 95%
1-(1,1-Difluoroethyl)-4-fluorobenzene
Formule: C₈H₇F₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11845816
CAS nummer: 55805-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR