3-(1,1-Difluoroethyl)pyridine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC520688-1G PC520688-500MG PC520688-5G
APOSPC520688-1GEA 399 EUR
APOSPC520688-1G APOSPC520688-500MG APOSPC520688-5G
3-(1,1-Difluoroethyl)pyridine 97%
3-(1,1-Difluoroethyl)pyridine
Formule: C₇H₇F₂N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12404951
CAS nummer: 114468-03-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR