3-Bromo-5-(trifluoromethyl)phenylacetic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC520693-1G PC520693-5G
APOSPC520693-1GEA 42.4 EUR
APOSPC520693-1G APOSPC520693-5G
3-Bromo-5-(trifluoromethyl)phenylacetic acid 97%
3-Bromo-5-(trifluoromethyl)phenylacetic acid
Formule: C9H6BrF3O2
MW: 283.04 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13194218
CAS nummer: 1161362-01-1
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR