3-(Trifluoromethyl)pyridazine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC520717-10G PC520717-1G PC520717-5G
APOSPC520717-10GEA 1540 EUR
APOSPC520717-10G APOSPC520717-1G APOSPC520717-5G
3-(Trifluoromethyl)pyridazine 97%
3-(Trifluoromethyl)pyridazine
Formule: C₅H₃F₃N₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16140190
CAS nummer: 132537-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR