1-(Bromomethyl)-4-(1,1,2,2-tetrafluoroethoxy)benzene 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC520733-25G PC520733-50G
APOSPC520733-25GEA 609 EUR
APOSPC520733-25G APOSPC520733-50G
1-(Bromomethyl)-4-(1,1,2,2-tetrafluoroethoxy)benzene 95%
1-(Bromomethyl)-4-(1,1,2,2-tetrafluoroethoxy)benzene
Formule: C₉H₇BrF₄O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16619616
CAS nummer: 67033-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR