2-Chloro-6-iodo-4-(trifluoromethyl)aniline 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC520752-10G PC520752-1G PC520752-5G
APOSPC520752-10GEA 1050 EUR
APOSPC520752-10G APOSPC520752-1G APOSPC520752-5G
2-Chloro-6-iodo-4-(trifluoromethyl)aniline 97%
2-Chloro-6-iodo-4-(trifluoromethyl)aniline
Formule: C₇H₄ClF₃IN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD20492612
CAS nummer: 1065102-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR