2,4-Difluoro-5-iodophenol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC52128-1G PC52128-5G
APOSPC52128-1GEA 281 EUR
APOSPC52128-1G APOSPC52128-5G
2,4-Difluoro-5-iodophenol
2,4-Difluoro-5-iodophenol
Formule: C₆H₃F₂IO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28986198
CAS nummer: 1935521-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR