1,1,1,3,3,3-Hexafluoro-2-(3-iodophenyl)propan-2-ol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC52192-1G PC52192-250MG
APOSPC52192-1GEA 176 EUR
APOSPC52192-1G APOSPC52192-250MG
1,1,1,3,3,3-Hexafluoro-2-(3-iodophenyl)propan-2-ol 95%
1,1,1,3,3,3-Hexafluoro-2-(3-iodophenyl)propan-2-ol
Formule: C₉H₅F₆IO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD31922516
CAS nummer: 2366994-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR