4-Bromo-2-chloro-3-fluorobenzyl alcohol 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC52203-1G PC52203-250MG PC52203-5G
APOSPC52203-1GEA 419 EUR
APOSPC52203-1G APOSPC52203-250MG APOSPC52203-5G
4-Bromo-2-chloro-3-fluorobenzyl alcohol 95%
4-Bromo-2-chloro-3-fluorobenzyl alcohol
Formule: C₇H₅BrClFO
Bewaar temperatuur: Koelkast
CAS nummer: 1891698-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR