2-Nitro-6-(trifluoromethyl)aniline 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5277-1G PC5277-5G PC5277-25G
APOSPC5277-1GEA 67.2 EUR
APOSPC5277-1G APOSPC5277-5G APOSPC5277-25G
2-Nitro-6-(trifluoromethyl)aniline 99%
2-Nitro-6-(trifluoromethyl)aniline
Formule: C₇H₅F₃N₂O₂
Smeltpunt: 48…52 °C
Vlampunt: >110 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08532494
CAS nummer: 24821-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR