4-Fluoro-DL-homophenylalanine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC530008-1G PC530008-250MG PC530008-500MG
APOSPC530008-1GEA 658 EUR
APOSPC530008-1G APOSPC530008-250MG APOSPC530008-500MG
4-Fluoro-DL-homophenylalanine 97%
4-Fluoro-DL-homophenylalanine
Formule: C₁₀H₁₂FNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD07372156
CAS nummer: 225233-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR