5-Bromo-2-fluoro-4-methylphenyl trifluoromethanesulphonate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC53047-1G PC53047-5G
APOSPC53047-1GEA 190 EUR
APOSPC53047-1G APOSPC53047-5G
5-Bromo-2-fluoro-4-methylphenyl trifluoromethanesulphonate
5-Bromo-2-fluoro-4-methylphenyl trifluoromethanesulphonate
Formule: C₈H₅BrF₄O₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28963743
CAS nummer: 1936290-01-5
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR