2-Fluoro-5-(hydroxymethyl)phenol 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC53135-1G PC53135-250MG PC53135-5G
APOSPC53135-1GEA 185 EUR
APOSPC53135-1G APOSPC53135-250MG APOSPC53135-5G
2-Fluoro-5-(hydroxymethyl)phenol 95+%
2-Fluoro-5-(hydroxymethyl)phenol
Formule: C₇H₇FO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD12031832
CAS nummer: 934241-78-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR