2-(2-Fluoro-6-nitrophenyl)acetic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC53138-1G PC53138-250MG
APOSPC53138-1GEA 191 EUR
APOSPC53138-1G APOSPC53138-250MG
2-(2-Fluoro-6-nitrophenyl)acetic acid
2-(2-Fluoro-6-nitrophenyl)acetic acid
Formule: C₈H₆FNO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD13185636
CAS nummer: 136916-19-3

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR