1-Butyl-3-methylimidazoliumtetrafluoroborate 98+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC53329-100G PC53329-25G
APOSPC53329-100GEA 187 EUR
APOSPC53329-100G APOSPC53329-25G
1-Butyl-3-methylimidazoliumtetrafluoroborate 98+%
1-Butyl-3-methylimidazoliumtetrafluoroborate
Formule: C₈H₁₅BF₄N₂
Smeltpunt: –71 °C
Dichtheid: 1,2077 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03095449
CAS nummer: 174501-65-6
UN: 3082
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR