5-Fluoro-2-iodophenol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC53345-1G PC53345-250MG PC53345-5G
APOSPC53345-1GEA 308 EUR
APOSPC53345-1G APOSPC53345-250MG APOSPC53345-5G
5-Fluoro-2-iodophenol
5-Fluoro-2-iodophenol
Formule: C₆H₄FIO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD16999912
CAS nummer: 186589-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR