4'-(1,1-Difluoroethyl)-2,2,2-trifluoroacetophenone

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC53356-1G PC53356-250MG
APOSPC53356-1GEA 366 EUR
APOSPC53356-1G APOSPC53356-250MG
4'-(1,1-Difluoroethyl)-2,2,2-trifluoroacetophenone
4'-(1,1-Difluoroethyl)-2,2,2-trifluoroacetophenone
Formule: C₁₀H₇F₅O
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD30184635
CAS nummer: 2091735-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR