4-Chloro-3-fluoro-2-methoxypyridine 95+%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC535045-100MG PC535045-250MG
APOSPC535045-100MGEA 78.9 EUR
APOSPC535045-100MG APOSPC535045-250MG
4-Chloro-3-fluoro-2-methoxypyridine 95+%
4-Chloro-3-fluoro-2-methoxypyridine
Formule: C₆H₅ClFNO
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD16610381
CAS nummer: 1227572-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR