3-Fluoro-5-(hydroxymethyl)benzeneboronic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC53505-1G PC53505-250MG PC53505-5G
APOSPC53505-1GEA 180 EUR
APOSPC53505-1G APOSPC53505-250MG APOSPC53505-5G
3-Fluoro-5-(hydroxymethyl)benzeneboronic acid
3-Fluoro-5-(hydroxymethyl)benzeneboronic acid
Formule: C₇H₈BFO₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD19687060
CAS nummer: 1146614-40-5

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR