5-Chloro-4-fluoropyridin-2-amine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC535063-1G PC535063-250MG
APOSPC535063-1GEA 163 EUR
APOSPC535063-1G APOSPC535063-250MG
5-Chloro-4-fluoropyridin-2-amine 97%
5-Chloro-4-fluoropyridin-2-amine
Formule: C₅H₄ClFN₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD22547000
CAS nummer: 1393574-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR