3-(2,2,2-Trifluoroethoxy)benzenesulphonyl chloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC53527-100MG PC53527-250MG PC53527-500MG PC53527-25MG
APOSPC53527-100MGEA 266 EUR
APOSPC53527-100MG APOSPC53527-250MG APOSPC53527-500MG APOSPC53527-25MG
3-(2,2,2-Trifluoroethoxy)benzenesulphonyl chloride
3-(2,2,2-Trifluoroethoxy)benzenesulphonyl chloride
Formule: C₈H₆ClF₃O₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD21601592
CAS nummer: 1365941-75-8
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR