3-[(2,2,3,3,4,4,5,5-Octafluoropent-1-yl)oxy]-1,2-propenoxide 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5353D-25G PC5353D-5G PC5353D-1G
APOSPC5353D-25GEA 442 EUR
APOSPC5353D-25G APOSPC5353D-5G APOSPC5353D-1G
3-[(2,2,3,3,4,4,5,5-Octafluoropent-1-yl)oxy]-1,2-propenoxide 97%
3-[(2,2,3,3,4,4,5,5-Octafluoropent-1-yl)oxy]-1,2-propenoxide
Formule: C₈H₈F₈O₂
Kookpunt: 75…79 °C (4 mm Hg)
Dichtheid: 1,509 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00054692
CAS nummer: 19932-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR