Ethyl 2-(4-Chlorophenyl)-2,2-difluoroacetate

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC53584-1G PC53584-250MG
APOSPC53584-1GEA 116 EUR
APOSPC53584-1G APOSPC53584-250MG
Ethyl 2-(4-Chlorophenyl)-2,2-difluoroacetate
Ethyl 2-(4-Chlorophenyl)-2,2-difluoroacetate
Formule: C₁₀H₉ClF₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11007726
CAS nummer: 130754-19-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR