Palladium(II) trifluoroacetate 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Palladium(II) 2,2,2-trifluoroacetate

PC5367-1G PC5367-5G
APOSPC5367-1GEA 103 EUR
APOSPC5367-1G APOSPC5367-5G
Palladium(II) trifluoroacetate 98%
Palladium(II) trifluoroacetate
Formule: C₄F₆O₄Pd
MW: 332.45 g/mol
Smeltpunt: 210 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013204
CAS nummer: 42196-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR