2-Amino-3-fluoro-5-(trifluoromethyl)pyridine 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5457-1G PC5457-5G PC5457-25G
APOSPC5457-1GEA 22 EUR
APOSPC5457-1G APOSPC5457-5G APOSPC5457-25G
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine 97%
2-Amino-3-fluoro-5-(trifluoromethyl)pyridine
Formule: C₆H₄F₄N₂
Smeltpunt: 64…68 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD10699117
CAS nummer: 852062-17-0
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR