6-Fluoro-4-hydrazinoquinoline hydrochloride

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5461-250MG PC5461-100MG PC5461-1G
APOSPC5461-250MGEA 155 EUR
APOSPC5461-250MG APOSPC5461-100MG APOSPC5461-1G
6-Fluoro-4-hydrazinoquinoline hydrochloride
6-Fluoro-4-hydrazinoquinoline hydrochloride
Formule: C₉H₈FN₃·ClH
Smeltpunt: >280 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02684141
CAS nummer: 1172049-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR