2,6-Difluoro-4-(trifluoromethoxy)aniline 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC55523-1G PC55523-250MG PC55523-5G
APOSPC55523-1GEA 579 EUR
APOSPC55523-1G APOSPC55523-250MG APOSPC55523-5G
2,6-Difluoro-4-(trifluoromethoxy)aniline 95%
2,6-Difluoro-4-(trifluoromethoxy)aniline
Formule: C₇H₄F₅NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28790978
CAS nummer: 1100833-35-9
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR