5-Fluoro-2-(trifluoromethyl)benzonitrile

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC5609-1G PC5609-500MG PC5609-5G PC5609-25G
APOSPC5609-1GEA 22 EUR
APOSPC5609-1G APOSPC5609-500MG APOSPC5609-5G APOSPC5609-25G
5-Fluoro-2-(trifluoromethyl)benzonitrile
5-Fluoro-2-(trifluoromethyl)benzonitrile
Formule: C₈H₃F₄N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00236281
CAS nummer: 240800-45-7
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR