2-Fluoro-6-methyl-4-nitroaniline 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC56269-1G PC56269-250MG PC56269-5G
APOSPC56269-1GEA 526 EUR
APOSPC56269-1G APOSPC56269-250MG APOSPC56269-5G
2-Fluoro-6-methyl-4-nitroaniline 95%
2-Fluoro-6-methyl-4-nitroaniline
Formule: C₇H₇FN₂O₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD28130101
CAS nummer: 1426805-91-5
UN: 2811
ADR: 6.1,III

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR