3-Amino-4-methoxyphenylsulphonyl fluoride 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC5637-10G PC5637-50G PC5637-1G
APOSPC5637-10GEA 0 EUR
APOSPC5637-10G APOSPC5637-50G APOSPC5637-1G
3-Amino-4-methoxyphenylsulphonyl fluoride 97%
3-Amino-4-methoxyphenylsulphonyl fluoride
Formule: C₇H₈FNO₃S
Smeltpunt: 60…63 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007417
CAS nummer: 498-74-8
EINECS: 207-870-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR