4-Bromo-2,5-bis(trifluoromethyl)benzyl bromide ≥95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC56686-250MG PC56686-1G
APOSPC56686-250MGEA 220 EUR
APOSPC56686-250MG APOSPC56686-1G
4-Bromo-2,5-bis(trifluoromethyl)benzyl bromide ≥95%
4-Bromo-2,5-bis(trifluoromethyl)benzyl bromide
Formule: C₉H₄Br₂F₆
MDL Nummer: MFCD28728007
CAS nummer: 1805513-09-0
UN: 3261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR