Cobalt(III) fluoride 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC5678-100G PC5678-25G PC5678-5G
APOSPC5678-100GEA 228 EUR
APOSPC5678-100G APOSPC5678-25G APOSPC5678-5G
Cobalt(III) fluoride 99%
Cobalt(III) fluoride

Hard fluorinating agent. Replaces organic hydrogen with fluorine, J. Appl. Chem., 1952, 127.

Formule: CoF₃
Dichtheid: 3,88 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010942
CAS nummer: 10026-18-3
EINECS: 233-062-4
UN: 3084
ADR: 8+5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR