2,3-Difluoro-5-methylbenzonitrile 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC56801-1G PC56801-500MG
APOSPC56801-1GEA 555 EUR
APOSPC56801-1G APOSPC56801-500MG
2,3-Difluoro-5-methylbenzonitrile 95%
2,3-Difluoro-5-methylbenzonitrile
Formule: C₈H₅F₂N
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD09996953
CAS nummer: 1003712-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR