3-Bromo-5-fluorophenylacetylene 96%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC56839-1G PC56839-250MG PC56839-5G
APOSPC56839-1GEA 187 EUR
APOSPC56839-1G APOSPC56839-250MG APOSPC56839-5G
3-Bromo-5-fluorophenylacetylene 96%
3-Bromo-5-fluorophenylacetylene
Formule: C₈H₄BrF
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD31654732
CAS nummer: 1935278-77-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR