2-Chloro-4-(trifluoromethyl)benzylamine 95%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC56851-1G PC56851-250MG
APOSPC56851-1GEA 1180 EUR
APOSPC56851-1G APOSPC56851-250MG
2-Chloro-4-(trifluoromethyl)benzylamine 95%
2-Chloro-4-(trifluoromethyl)benzylamine
Formule: C₈H₇ClF₃N
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06212820
CAS nummer: 581813-20-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR