4-[(4-Fluorophenyl)-hydroxymethyl]benzonitrile 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC56868-1G PC56868-25G PC56868-5G
APOSPC56868-1GEA 93.5 EUR
APOSPC56868-1G APOSPC56868-25G APOSPC56868-5G
4-[(4-Fluorophenyl)-hydroxymethyl]benzonitrile 98%
4-[(4-Fluorophenyl)-hydroxymethyl]benzonitrile
Formule: C₁₄H₁₀FNO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11096849
CAS nummer: 220583-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR