6-Bromo-3-chloro-2-fluorobenzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC57054-1G PC57054-250MG PC57054-5G
APOSPC57054-1GEA 46.8 EUR
APOSPC57054-1G APOSPC57054-250MG APOSPC57054-5G
6-Bromo-3-chloro-2-fluorobenzaldehyde
6-Bromo-3-chloro-2-fluorobenzaldehyde
Formule: C₇H₃BrClFO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD11520661
CAS nummer: 1114809-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR