2-Fluoro-4-(trifluoromethoxy)benzyl alcohol 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC57059-1G PC57059-5G PC57059-10G
APOSPC57059-1GEA 163 EUR
APOSPC57059-1G APOSPC57059-5G APOSPC57059-10G
2-Fluoro-4-(trifluoromethoxy)benzyl alcohol 98%
2-Fluoro-4-(trifluoromethoxy)benzyl alcohol
Formule: C₈H₆F₄O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD16652441
CAS nummer: 1240257-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR