(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid 97%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PC5719-1G
APOSPC5719-1GEA 132 EUR
APOSPC5719-1G
(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid 97%
(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid
Formule: C₅H₈FNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02683983
CAS nummer: 2438-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR