2,4-Difluoro-3-(trifluoromethoxy)phenol

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC57486-1G PC57486-5G
APOSPC57486-1GEA 577 EUR
APOSPC57486-1G APOSPC57486-5G
2,4-Difluoro-3-(trifluoromethoxy)phenol
2,4-Difluoro-3-(trifluoromethoxy)phenol
Formule: C₇H₃F₅O₂
MDL Nummer: MFCD34595684
CAS nummer: 2383762-98-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR