2-(Bromomethyl)-7-(trifluoromethyl)benzothiophene

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC57778-1G PC57778-250MG
APOSPC57778-1GEA 125 EUR
APOSPC57778-1G APOSPC57778-250MG
2-(Bromomethyl)-7-(trifluoromethyl)benzothiophene
2-(Bromomethyl)-7-(trifluoromethyl)benzothiophene
Formule: C₁₀H₆BrF₃S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD28399009
CAS nummer: 1858242-05-3
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR