2-Bromo-4-(trifluoromethyl)phenylacetic acid

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5830-1G PC5830-25G PC5830-5G
APOSPC5830-1GEA 61.4 EUR
APOSPC5830-1G APOSPC5830-25G APOSPC5830-5G
2-Bromo-4-(trifluoromethyl)phenylacetic acid
2-Bromo-4-(trifluoromethyl)phenylacetic acid
Formule: C₉H₆BrF₃O₂
Bewaar temperatuur: Kamertemperatuur
CAS nummer: 518070-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR