2-Chloro-6-(trifluoromethyl)nicotinamide 98%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

PC5877-1G PC5877-5G PC5877-25G
APOSPC5877-1GEA 26.3 EUR
APOSPC5877-1G APOSPC5877-5G APOSPC5877-25G
2-Chloro-6-(trifluoromethyl)nicotinamide 98%
2-Chloro-6-(trifluoromethyl)nicotinamide
Formule: C₇H₄ClF₃N₂O
Smeltpunt: 198…200 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01862659
CAS nummer: 386704-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR