2-Fluoro-5-(trifluoromethoxy)benzylamine

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC5899-10G PC5899-1G PC5899-5G
APOSPC5899-10GEA 646 EUR
APOSPC5899-10G APOSPC5899-1G APOSPC5899-5G
2-Fluoro-5-(trifluoromethoxy)benzylamine
2-Fluoro-5-(trifluoromethoxy)benzylamine
Formule: C₈H₇F₄NO
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04115895
CAS nummer: 886498-13-1
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR