2-Fluoro-5-(trifluoromethoxy)aniline 99%

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

PC5900-1G PC5900-25G PC5900-5G
APOSPC5900-1GEA 30.8 EUR
APOSPC5900-1G APOSPC5900-25G APOSPC5900-5G
2-Fluoro-5-(trifluoromethoxy)aniline 99%
2-Fluoro-5-(trifluoromethoxy)aniline
Formule: C₇H₅F₄NO
Kookpunt: 64…65 °C/15mmHg
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD09832254
CAS nummer: 116369-23-4
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR